Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-chloro-5-fluorophenol ≥97% research grade

Supplier: AOB CHEM USA
Warning

58292-1G 58292-25G 58292-500MG 58292-5G
76905-286EA 188.85 USD InStock
76905-286 76905-288 76905-290 76905-292
2-Bromo-4-chloro-5-fluorophenol ≥97% research grade
2-Bromo-4-chloro-5-fluorophenol


Formula: C₆H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD26895908
CAS Number: 157165-23-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty