Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-iodoisonicotinic acid ≥97%

Supplier: AOB CHEM USA
Warning

60718-1G 60718-25G 60718-500MG 60718-5G
76905-424EA 164.44 USD InStock
76905-424 76905-426 76905-428 76905-430
2,6-Dichloro-3-iodoisonicotinic acid ≥97%
2,6-Dichloro-3-iodoisonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₂Cl₂INO₂
Storage Temperature: Ambient
CAS Number: 2386622-46-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty