Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-bromo-4-methylpicolinate ≥97% research grade

Supplier: AOB CHEM USA
Warning

24323-1G 24323-25G 24323-5G
76928-340EA 112.46 USD InStock
76928-340 76928-342 76928-344
Methyl-5-bromo-4-methylpicolinate ≥97% research grade
Methyl-5-bromo-4-methylpicolinate


Formula: C₈H₈BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07375102
CAS Number: 886365-06-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty