Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloro-5-(hydroxymethyl)phenyl)boronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

31164-1G 31164-25G 31164-5G
76930-598EA 64.72 USD InStock
76930-598 76930-600 76930-602
(3-Chloro-5-(hydroxymethyl)phenyl)boronic acid ≥95% research grade
(3-Chloro-5-(hydroxymethyl)phenyl)boronic acid


Formula: C₇H₈BClO₃
Storage Temperature: Ambient
MDL Number: MFCD22397986
CAS Number: 2121512-87-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty