Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxycarbonyl-3-nitrophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

40170-1G 40170-25G 40170-5G
76932-352EA 96.55 USD InStock
76932-352 76932-354 76932-356
4-Methoxycarbonyl-3-nitrophenylboronic acid ≥97%
4-Methoxycarbonyl-3-nitrophenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₈BNO₆
Storage Temperature: Ambient
MDL Number: MFCD12032027
CAS Number: 85107-56-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty