Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-chloro-5-methylnicotinate ≥97%

Supplier: AOB CHEM USA
Warning

21585-1G 21585-25G 21585-500MG 21585-5G
76937-936EA 155.96 USD InStock
76937-936 76937-938 76937-940 76937-942
Methyl 6-chloro-5-methylnicotinate ≥97%
Methyl 6-chloro-5-methylnicotinate
Precaución: for Research Use Only


Formula: C₈H₈ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18258043
CAS Number: 65169-42-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty