Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,6-dibromonicotinate ≥97%

Supplier: AOB CHEM USA
Warning

21762-1G 21762-25G 21762-500MG 21762-5G
76938-004EA 259.93 USD InStock
76938-004 76938-006 76938-008 76938-010
Methyl-2,6-dibromonicotinate ≥97%
Methyl-2,6-dibromonicotinate
Precaución: for Research Use Only


Formula: C₇H₅Br₂NO₂
Storage Temperature: Ambient
CAS Number: 1009735-22-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty