Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-chloro-2,4-dimethoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

32341-1G 32341-25G 32341-500MG 32341-5G
76940-194EA 292.81 USD InStock
76940-194 76940-196 76940-198 76940-200
Methyl 5-chloro-2,4-dimethoxybenzoate ≥97%
Methyl 5-chloro-2,4-dimethoxybenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₁ClO₄
Storage Temperature: Ambient
MDL Number: MFCD24108547
CAS Number: 174261-01-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty