Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,4,6-trichlorobenzoate ≥95%

Supplier: AOB CHEM USA
Warning

49843-1G 49843-25G 49843-500MG 49843-5G
76942-904EA 263.11 USD InStock
76942-904 76942-906 76942-908 76942-910
Ethyl 2,4,6-trichlorobenzoate ≥95%
Ethyl 2,4,6-trichlorobenzoate


Formula: C₉H₇Cl₃O₂
Storage Temperature: Ambient
MDL Number: MFCD12024964
CAS Number: 73981-96-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty