Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-iodophenol ≥97% research grade

Supplier: AOB CHEM USA
Warning

28307-1G 28307-25G 28307-500MG 28307-5G
76900-938EA 153.84 USD InStock
76900-938 76900-940 76900-942 76900-944
2-Chloro-6-iodophenol ≥97% research grade
2-Chloro-6-iodophenol


Formula: C₆H₄ClIO
Storage Temperature: Refrigerator
MDL Number: MFCD15833001
CAS Number: 28177-52-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty