Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyloxy-4-bromo-2-methylbenzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

11206-1G 11206-25G 11206-500MG 11206-5G
76882-156EA 260.99 USD InStock
76882-156 76882-158 76882-160 76882-162
1-Benzyloxy-4-bromo-2-methylbenzene ≥97% research grade
1-Benzyloxy-4-bromo-2-methylbenzene


Formula: C₁₄H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD18361005
CAS Number: 338454-32-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty