Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloro-5-formylpyridine ≥95%

Supplier: AOB CHEM USA
Warning

21284-1G 21284-25G 21284-500MG 21284-5G
76884-772EA 142.17 USD InStock
76884-772 76884-774 76884-776 76884-778
2,3-Dichloro-5-formylpyridine ≥95%
2,3-Dichloro-5-formylpyridine


Formula: C₆H₃Cl₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD08448226
CAS Number: 71690-05-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty