Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloro-2-fluoro-4-methylphenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

44698-1G 44698-25G 44698-500MG 44698-5G
76889-900EA 164.44 USD InStock
76889-900 76889-902 76889-904 76889-906
(3-Chloro-2-fluoro-4-methylphenyl)boronic acid ≥97% research grade
(3-Chloro-2-fluoro-4-methylphenyl)boronic acid


Formula: C₇H₇BClFO₂
Storage Temperature: Ambient
MDL Number: MFCD23712893
CAS Number: 2377608-77-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty