Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

10165-1G 10165-25G 10165-500MG 10165-5G
76906-296EA 260.99 USD InStock
76906-296 76906-298 76906-300 76906-302
4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester ≥97%
4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₇H₂₀BClN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD06798267
CAS Number: 1073354-22-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty