Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dfluoro-2-methoxybenzoic acid methyl ester ≥97%

Supplier: AOB CHEM USA
Warning

23359-1G 23359-25G 23359-500MG 23359-5G
76911-226EA 194.15 USD InStock
76911-226 76911-228 76911-230 76911-232
4,5-Dfluoro-2-methoxybenzoic acid methyl ester ≥97%
4,5-Dfluoro-2-methoxybenzoic acid methyl ester


Formula: C₉H₈F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD10566200
CAS Number: 1261444-15-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty