Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-6-isopropyl-3-pyridylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

16846-1G 16846-25G 16846-500MG 16846-5G
76896-610EA 266.29 USD InStock
76896-610 76896-612 76896-614 76896-616
2-Methoxy-6-isopropyl-3-pyridylboronic acid ≥97%
2-Methoxy-6-isopropyl-3-pyridylboronic acid


Formula: C₉H₁₄BNO₃
Storage Temperature: Ambient
MDL Number: MFCD16295243
CAS Number: 477598-24-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty