Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-bromobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

20203-1G 20203-25G 20203-500MG 20203-5G
76897-816EA 292.81 USD InStock
76897-816 76897-818 76897-820 76897-822
2,6-Dichloro-3-bromobenzoic acid ≥97% research grade
2,6-Dichloro-3-bromobenzoic acid


Formula: C₇H₃BrCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18432783
CAS Number: 80257-12-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty