Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropyl-4-methoxyphenol ≥97%

Supplier: AOB CHEM USA
Warning

33150-1G 33150-25G 33150-500MG 33150-5G
76902-454EA 245.07 USD InStock
76902-454 76902-456 76902-458 76902-460
2-Isopropyl-4-methoxyphenol ≥97%
2-Isopropyl-4-methoxyphenol


Formula: C₁₀H₁₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07787576
CAS Number: 13522-86-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty