Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methoxyisonicotinic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

15781-1G 15781-25G 15781-5G
76925-538EA 62.6 USD InStock
76925-538 76925-540 76925-542
5-Bromo-2-methoxyisonicotinic acid ≥95% research grade
5-Bromo-2-methoxyisonicotinic acid


Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375111
CAS Number: 886365-22-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty