Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

19307-1G 19307-25G 19307-5G
76926-738EA 73.21 USD InStock
76926-738 76926-740 76926-742
5-Chloro-2-formylphenylboronic acid pinacol ester ≥97%
5-Chloro-2-formylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₆BClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16295261
CAS Number: 1246633-36-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty