Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-isopropylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

19989-1G 19989-25G 19989-500MG 19989-5G
76926-940EA 171.87 USD InStock
76926-940 76926-942 76926-944 76926-946
4-Bromo-2-isopropylbenzoic acid ≥97%
4-Bromo-2-isopropylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19686186
CAS Number: 741698-83-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty