Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylaminomethylphenylboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

22779-1G 22779-25G 22779-500MG 22779-5G
76927-872EA 367.08 USD InStock
76927-872 76927-874 76927-876 76927-878
4-Methylaminomethylphenylboronic acid ≥95%
4-Methylaminomethylphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₁₂BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD03094948
CAS Number: 518336-26-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty