Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodobenzo[b]thiophene-2-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

99940-1G 99940-25G 99940-500MG 99940-5G
76936-156EA 162.32 USD InStock
76936-156 76936-158 76936-160 76936-162
4-Iodobenzo[b]thiophene-2-carboxylic acid ≥97%
4-Iodobenzo[b]thiophene-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₅IO₂S
Storage Temperature: Ambient
CAS Number: 176549-83-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty