Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-1,3-dihydro-2,1-benzoxaborol-1-ol ≥97%

Supplier: AOB CHEM USA
Warning

16684-1G 16684-500MG 16684-5G
76937-146EA 468.92 USD InStock
76937-146 76937-148 76937-150
6-Chloro-1,3-dihydro-2,1-benzoxaborol-1-ol ≥97%
6-Chloro-1,3-dihydro-2,1-benzoxaborol-1-ol
Precaución: for Research Use Only


Formula: C₇H₆BClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD22415418
CAS Number: 174671-43-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty