tert-Butyl 4-(trifluoromethyl)benzoate ≥95%
Supplier: AOB CHEM USA
|
Warning
|
28121-1G
28121-25G
28121-500MG
28121-5G
76939-300EA
369.2
USD
76939-300
76939-302
76939-304
76939-306
tert-Butyl 4-(trifluoromethyl)benzoate ≥95%
tert-Butyl 4-(trifluoromethyl)benzoate
Formula:
C₁₂H₁₃F₃O₂ MW: 246.226 g/mol Storage Temperature: Ambient |
CAS Number:
196934-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...