Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[2-Methyl-5-(4,4,5,5-Tetramethyl[1,3,2]Dioxaborolan-2-Yl)Phenyl]Methanesulfonamide ≥97%

Supplier: AOB CHEM USA
Warning

34597-1G 34597-25G 34597-500MG 34597-5G
76940-872EA 409.51 USD InStock
76940-872 76940-874 76940-876 76940-878
N-[2-Methyl-5-(4,4,5,5-Tetramethyl[1,3,2]Dioxaborolan-2-Yl)Phenyl]Methanesulfonamide ≥97%
N-[2-Methyl-5-(4,4,5,5-Tetramethyl[1,3,2]Dioxaborolan-2-Yl)Phenyl]Methanesulfonamide
Precaución: for Research Use Only


Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty