Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-bromo-3,5-dichlorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

50036-1G 50036-25G 50036-5G
76942-934EA 81.69 USD InStock
76942-934 76942-936 76942-938
Ethyl 4-bromo-3,5-dichlorobenzoate ≥97%
Ethyl 4-bromo-3,5-dichlorobenzoate


Formula: C₉H₇BrCl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD14704249
CAS Number: 1239741-87-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty