Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-Neomenthyldiphenylphosphine 98%

Supplier: AMBEED, INC
Warning

A359966-100MG A359966-1G A359966-250MG
76978-574EA 66 USD InStock
76978-574 76978-576 76978-578
(S)-(+)-Neomenthyldiphenylphosphine 98%
(S)-(+)-Neomenthyldiphenylphosphine
Precaución: for Research Use Only


Formula: C₂₂H₂₉P
MDL Number: MFCD00013415
CAS Number: 43077-29-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty