Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥97%, ee 99%

Supplier: AMBEED, INC
Warning

A465873-1G A465873-5G A465873-25G
76979-602EA 10 USD InStock
76979-602 76979-604 77594-466
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥97%, ee 99%
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
Precaución: for Research Use Only


Formula: C₂₄H₂₂O₄
MDL Number: MFCD03788935
CAS Number: 173831-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty