Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-2,3,6-trimethylbenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A279974-250MG A279974-1G A279974-25G
76977-384EA 5 USD InStock
76977-384 77672-438 77672-440
4-Methoxy-2,3,6-trimethylbenzaldehyde 95%
4-Methoxy-2,3,6-trimethylbenzaldehyde
Precaución: For research use only.


Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00456729
CAS Number: 54344-92-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty