Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitro-o-cresol 98%

Supplier: AMBEED, INC
Warning

A267235-10G A267235-1G A267235-25G A267235-5G
76977-122EA 167.63 USD InStock
76977-122 76977-124 76977-126 76977-128 76977-130
3-Nitro-o-cresol 98%
3-Nitro-o-cresol
Precaución: for Research Use Only


Formula: C₇H₇NO₃
MDL Number: MFCD00007241
CAS Number: 5460-31-1
UN: 2446
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty