Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-(m-tolyl)propanoic acid 97%

Supplier: AMBEED, INC
Warning

A235511-10G A235511-1G A235511-250MG A235511-25G A235511-5G
76976-322EA 86 USD InStock
76976-322 76976-324 76976-326 76976-328 76976-330
2-Amino-3-(m-tolyl)propanoic acid 97%
2-Amino-3-(m-tolyl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂
MDL Number: MFCD05227937
CAS Number: 5472-70-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty