Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodoadenosine 97%

Supplier: AMBEED, INC
Warning

A148248-1G A148248-250MG A148248-100MG
76972-948EA 61 USD InStock
76972-948 76972-950 77671-084
2-Iodoadenosine 97%
2-Iodoadenosine
Precaución: For research use only.


Formula: C₁₀H₁₂IN₅O₄
MDL Number: MFCD01320413
CAS Number: 35109-88-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty