4-Iodophthalonitrile 95%

Supplier: AMBEED, INC
Warning

A469811-100MG A469811-1G A469811-250MG A469811-5G
76979-644EA 10 USD
76979-644 76979-646 76979-648 77673-208
4-Iodophthalonitrile 95%
4-Iodophthalonitrile

Precaución: for Research Use Only

Formula: C₈H₃IN₂
MW: 254.03
MDL Number: MFCD01863730
CAS Number: 69518-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR