Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,2,4]Triazolo[4,3-a]pyridin-6-amine 95%

Supplier: AMBEED, INC
Warning

A107576-100MG A107576-1G A107576-250MG A107576-5G
76970-510EA 62 USD InStock
76970-510 76970-512 76970-514 76970-516
[1,2,4]Triazolo[4,3-a]pyridin-6-amine 95%
[1,2,4]Triazolo[4,3-a]pyridin-6-amine
Precaución: for Research Use Only


Formula: C₆H₆N₄
MDL Number: MFCD11597277
CAS Number: 1082448-58-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty