Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoroisoquinoline 95%

Supplier: AMBEED, INC
Warning

A231751-100MG A231751-10G A231751-1G A231751-250MG A231751-5G
76976-176EA 5 USD InStock
76976-176 76976-178 76976-180 76976-182 76976-184
6-Fluoroisoquinoline 95%
6-Fluoroisoquinoline
Precaución: for Research Use Only


Formula: C₉H₆FN
MDL Number: MFCD11226829
CAS Number: 1075-11-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty