Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-chloropicolinic acid 95%

Supplier: AMBEED, INC
Warning

A172558-100MG A172558-1G A172558-250MG
76974-078EA 35 USD InStock
76974-078 76974-080 76974-082
5-Bromo-6-chloropicolinic acid 95%
5-Bromo-6-chloropicolinic acid
Precaución: for Research Use Only


Formula: C₆H₃BrClNO₂
MDL Number: MFCD09910220
CAS Number: 959958-25-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty