Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-1H-pyrazol-5-ol 97%

Supplier: AMBEED, INC
Warning

A115969-100G A115969-10G A115969-25G A115969-5G
77026-094EA 168 USD InStock
77026-094 77026-096 77026-098 77026-100
3-Amino-1H-pyrazol-5-ol 97%
3-Amino-1H-pyrazol-5-ol
Precaución: for Research Use Only


Formula: C₃H₅N₃O
MDL Number: MFCD00233932
CAS Number: 53666-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty