Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-(4-formylphenyl)acrylate 98%

Supplier: AMBEED, INC
Warning

A140518-10G A140518-1G A140518-250MG A140518-25G A140518-5G
77027-498EA 75 USD InStock
77027-498 77027-500 77027-502 77027-504 77027-506
Methyl 3-(4-formylphenyl)acrylate 98%
Methyl 3-(4-formylphenyl)acrylate
Precaución: for Research Use Only


Formula: C₁₁H₁₀O₃
MDL Number: MFCD00157179
CAS Number: 58045-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty