1-Methylhydrazinecarbothioamide 97%

Supplier: AMBEED, INC
Warning

A161795-100G A161795-1G A161795-25G A161795-5G
76997-830EA 359 USD
76997-830 76997-832 76997-834 76997-836
1-Methylhydrazinecarbothioamide 97%
1-Methylhydrazinecarbothioamide

Precaución: for Research Use Only

Formula: C₂H₇N₃S
MW: 105.16 g/mol
MDL Number: MFCD00010147
CAS Number: 6938-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR