Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-(trifluoromethyl)imidazo[1,2-b]pyridazine 96%

Supplier: AMBEED, INC
Warning

A176819-100MG A176819-1G A176819-250MG
77011-814EA 11 USD InStock
77011-814 77011-816 77011-818
3-Bromo-2-(trifluoromethyl)imidazo[1,2-b]pyridazine 96%
3-Bromo-2-(trifluoromethyl)imidazo[1,2-b]pyridazine
Precaución: for Research Use Only


Formula: C₇H₃BrF₃N₃
MDL Number: MFCD13193556
CAS Number: 1260817-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty