Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-iodo-7-azaindole 95%

Supplier: AMBEED, INC
Warning

A184110-1G A184110-250MG A184110-5G
77029-566EA 51 USD InStock
77029-566 77029-568 77029-570
4-Bromo-3-iodo-7-azaindole 95%
4-Bromo-3-iodo-7-azaindole
Precaución: for Research Use Only


Formula: C₇H₄BrIN₂
MDL Number: MFCD09880123
CAS Number: 1000340-34-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty