Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Chloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98+%

Supplier: AMBEED, INC
Warning

A190485-100MG A190485-1G A190485-250MG A190485-5G
77012-296EA 18 USD InStock
77012-296 77012-298 77012-300 77012-302
2-(2-Chloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98+%
2-(2-Chloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₃H₁₈BClO₂
MDL Number: MFCD06659053
CAS Number: 1144097-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty