Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trityl candesartan 97%

Supplier: AMBEED, INC
Danger

A200091-1G A200091-250MG A200091-5G
76999-286EA 36 USD InStock
76999-286 76999-288 76999-290
Trityl candesartan 97%
Trityl candesartan
Precaución: for Research Use Only


Formula: C₄₃H₃₄N₆O₃
MDL Number: MFCD08436154
CAS Number: 139481-72-4
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty