3-Bromo-1-methyl-1H-indazole 98%

Supplier: AMBEED, INC
Danger

A210958-250MG A210958-5G
76986-716EA 26 USD
76986-716 76986-718
3-Bromo-1-methyl-1H-indazole 98%
3-Bromo-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0586 g/mol
MDL Number: MFCD12923368
CAS Number: 326474-67-3
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR