Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4,5-Tetramethylaniline 95%

Supplier: AMBEED, INC
Danger

A213463-100MG A213463-1G A213463-250MG
76986-804EA 165 USD InStock
76986-804 76986-806 76986-808
2,3,4,5-Tetramethylaniline 95%
2,3,4,5-Tetramethylaniline
Precaución: for Research Use Only


Formula: C₁₀H₁₅N
MDL Number: MFCD19203597
CAS Number: 2217-45-0
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty