Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5,6-dimethoxy-3-methylpyridine 95%

Supplier: AMBEED, INC
Warning

A214990-100MG A214990-1G A214990-5G
77013-074EA 63 USD InStock
77013-074 77013-076 77013-078 77013-080
2-Bromo-5,6-dimethoxy-3-methylpyridine 95%
2-Bromo-5,6-dimethoxy-3-methylpyridine
Precaución: for Research Use Only


Formula: C₈H₁₀BrNO₂
MDL Number: MFCD27996780
CAS Number: 64837-91-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty