Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-4-trifluoromethoxyphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A216685-100MG A216685-1G A216685-250MG
77013-128EA 39 USD InStock
77013-128 77013-130 77013-132
3-Methyl-4-trifluoromethoxyphenylboronic acid 97%
3-Methyl-4-trifluoromethoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₈BF₃O₃
MDL Number: MFCD11504858
CAS Number: 871362-79-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty