Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methylthio)pyrazine 98%

Supplier: AMBEED, INC
Danger

A236020-100G A236020-10G A236020-25G A236020-5G
77000-304EA 49 USD InStock
77000-304 77000-306 77000-308 77000-310
2-(Methylthio)pyrazine 98%
2-(Methylthio)pyrazine
Precaución: for Research Use Only


Formula: C₅H₆N₂S
MDL Number: MFCD00041024
CAS Number: 21948-70-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty