Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N'-Isopropylacetohydrazide 95%

Supplier: AMBEED, INC
Warning

A253096-100MG A253096-1G A253096-250MG
76987-956EA 70 USD InStock
76987-956 76987-958 76987-960
N'-Isopropylacetohydrazide 95%
N'-Isopropylacetohydrazide
Precaución: for Research Use Only


Formula: C₅H₁₂N₂O
MDL Number: MFCD01672496
CAS Number: 4466-50-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty